EMAIL THIS PAGE TO A FRIEND
Annals of hematology
Mutant DNMT3A clone evading chemotherapy and infiltrating central nervous system in a patient with molecularly good-risk acute myeloid leukemia.
PMID 24337413