EMAIL THIS PAGE TO A FRIEND
Muscle & nerve
Blood-based diagnostic testing for Pompe disease: consistency between GAA enzyme activity in dried blood spots and GAA gene sequencing results.
PMID 24338800