EMAIL THIS PAGE TO A FRIEND
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi
Importance of sulfonamide moiety in current and future therapy.
PMID 24340546
Abstract
Sulfonamides and their different derivatives are extensively used in therapy due to their pharmacological properties. Sulfa drugs were amongst the oldest synthesized antimicrobial agents and are still widely used today to treat different microbial infections. Clinical treatment with sulfonamides has regained confidence with the use of a combination of sulfamethoxazole and trimethoprim to treat urinary tract bacterial infections. Today, they are widely used as antimicrobial agents, chiefly because of their low cost, low toxicity and excellent activity against bacterial diseases. Over the course of time, the application of sulfonamides has been extended from their use as antimicrobial agents to anticancer agents, antiglaucoma agents, inhibitors of gamma-secretase, cyclooxgenase-2 and lipoxygenase, anticonvulsivant agents, hypoglycemic agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2487 Cotrimoxazole, Ready Made Solution, 100 mg/mL in DMSO
C14H18N4O3 · 5 C10H11N3O3S