EMAIL THIS PAGE TO A FRIEND
Genetic counseling (Geneva, Switzerland)
Crigler-Najjar syndrome type I in a Turkish newborn caused by a novel mutation and Gilbert type genetic defect.
PMID 24341141
Abstract
Crigler-Najjar syndrome (CNS), caused by deficiency of bilirubin uridine diphosphate glucuronosyltransferase (UGT) 1A1, is a rare and autosomal recessive inherited disorder characterized by severe unconjugated nonhemolytic hyperbilirubinemia since birth. We present a girl with CNS type I caused by a novel mutation and Gilbert type genetic defect. Gilbert's Syndrome (GS) and CNS type I both involve abnormalities in bilirubin conjugation secondary to deficiency of bilirubin UGT. The combined defects even in benign genetic forms were shown to cause more serious clinical disease. The patient has been treated with daily home-based phototherapy for more than nine months and considered as a candidate for liver transplantation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV44294
Anti-UGT1A1 antibody produced in rabbit, IgG fraction of antiserum