EMAIL THIS PAGE TO A FRIEND
Biomaterials
Drug-loaded gold plasmonic nanoparticles for treatment of multidrug resistance in cancer.
PMID 24342728
Abstract
To investigate the possibility of treating multidrug-resistant tumors with targeted chemo-photothermal treatment, we conducted in vitro and in vivo studies using a doxorubicin (DOX)-resistant DLD-1 cell line (DLD-1/DOX) and nude mice with human xenograft tumors, respectively. The chemo-photothermal treatment consisted of DOX-loaded-poly(lactic-co-glycolic acid)-Au half-shell nanoparticles with targeting moieties of anti-death receptor-4 monoclonal antibody conjugated to the Au surface. The cells or xenografted tumors treated with nanoparticles were exposed to near infrared light for 10 min, which caused an increase in temperature to 45 °C. Chemo-photothermal treatment resulted in a large reduction in the rate of tumor xenograft growth on DLD-1/DOX tumor-bearing mice with a much smaller dose of DOX than conventional DOX chemotherapy. These results demonstrate that targeted chemo-photothermal treatment can provide high therapeutic efficacy and low toxicity in the treatment of multidrug-resistant tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
DLD-1, 90102540