EMAIL THIS PAGE TO A FRIEND
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Enhancement of Achilles tendon repair mediated by matrix metalloproteinase inhibition via systemic administration of doxycycline.
PMID 24346815
Abstract
Collagenases or matrix metalloproteinases (MMPs) have been shown to play an important role in the matrix degradation cascade associated with Achilles tendon rupture and disease. The goal of this study was to examine the effects of daily administration of doxycycline (Doxy) through oral gavage on MMP activity and on the repair quality of Achilles tendons in vivo. Our findings indicate that Achilles tendon transection resulted in increasing MMP-8 activity from 2 to 6 weeks post-injury, with peak increases in activity occurring at 4 weeks post-injury. Doxy adiministration at clinically relevant serum concentrations was found to significantly inhibit MMP activity after continuous treatment for 4 weeks, but not for continuous administration for shorter durations (96 h or 2 weeks). Extended doxy administration was also associated with improved collagen fibril organization, and enhanced biomechanical properties (stiffness, ultimate tensile strength, maximum load to failure, and elastic toughness). Our findings indicate that a temporal delay exists between Achilles tendon transection and associated increases in MMP-8 activity in situ. Our findings suggest that inhibition of MMP-8 at its peak activity levels ameliorates fibrosis development and improves biomechanical properties of the Achilles tendon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3000000
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
1226003
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O