EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
A Cdc42- and Rac-interactive binding (CRIB) domain mediates functions of coronin.
PMID 24347642
Abstract
The Cdc42- and Rac-interactive binding motif (CRIB) of coronin binds to Rho GTPases with a preference for GDP-loaded Rac. Mutation of the Cdc42- and Rac-interactive binding motif abrogates Rac binding. This results in increased 1evels of activated Rac in coronin-deficient Dictyostelium cells (corA(-)), which impacts myosin II assembly. corA(-) cells show increased accumulation of myosin II in the cortex of growth-phase cells. Myosin II assembly is regulated by myosin heavy chain kinase-mediated phosphorylation of its tail. Kinase activity depends on the activation state of the p21-activated kinase a. The myosin II defect of corA(-) mutant is alleviated by dominant-negative p21-activated kinase a. It is rescued by wild-type coronin, whereas coronin carrying a mutated Cdc42- and Rac-interactive binding motif failed to rescue the myosin defect in corA(-) mutant cells. Ectopically expressed myosin heavy chain kinases affinity purified from corA(-) cells show reduced kinase activity. We propose that coronin through its affinity for GDP-Rac regulates the availability of GTP-Rac for activation of downstream effectors.