EMAIL THIS PAGE TO A FRIEND
Advanced healthcare materials
High-density polymer microarrays: identifying synthetic polymers that control human embryonic stem cell growth.
PMID 24353271
Abstract
The fabrication of high-density polymer microarray is described, allowing the simultaneous and efficient evaluation of more than 7000 different polymers in a single-cellular-based screen. These high-density polymer arrays are applied in the search for synthetic substrates for hESCs culture. Up-scaling of the identified hit polymers enables long-term cellular cultivation and promoted successful stem-cell maintenance.