EMAIL THIS PAGE TO A FRIEND
Human Vaccines & Immunotherapeutics
Yeast-expressed recombinant protein of the receptor-binding domain in SARS-CoV spike protein with deglycosylated forms as a SARS vaccine candidate.
PMID 24355931