EMAIL THIS PAGE TO A FRIEND
Dentistry today
Class II composite placement is difficult! Solutions to help overcome the clinical challenges.
PMID 24358548
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
494356
Bisphenol A glycerolate dimethacrylate, glycerol/phenol 1
C29H36O8