EMAIL THIS PAGE TO A FRIEND
Dentistry today
Total dental aesthetics.
PMID 24358552
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7630
Hyaluronic acid sodium salt from bovine vitreous humor
H5388
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
53747
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
40583
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
49775
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
63357
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
97616
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
94137
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
73641
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
41897
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
42686
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
08185
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
51967
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
96144
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
06742
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
00941
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
H9390
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide