EMAIL THIS PAGE TO A FRIEND
International journal of orthodontics (Milwaukee, Wis.)
Comparison between drill and drill free screws as a source of rigid orthodontic anchorage: a prospective clinical study.
PMID 24358660
Abstract
The study was undertaken to evaluate and compare the stability and surrounding soft tissue reaction of the Drill and Drill-free miniscrews when used as anchorage unit for en-masse retraction of maxillary anteriors. 20 patients were selected. Of the total 20 Drill and drill free screws, halfscrews were placed on the right side and half on the left side of maxilla and clinical evaluation for mobility and any surrounding soft tissue inflammation was done. There were no statistically significant differences with respect to Stability/Failure and soft tissue reaction for Drill and Drill-free screws when used as anchorage units.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
IRMM531A
Titanium, IRMM® certified Reference Material, 0.1 mm foil
Ti
IRMM531B
Titanium, IRMM® certified Reference Material, 0.5 mm foil
Ti
IRMM531C
Titanium, IRMM® certified Reference Material, 0.5 mm wire
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
348821
Titanium, foil, thickness 0.05 mm, 99.99% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
513415
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
348864
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
266019
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Ti