EMAIL THIS PAGE TO A FRIEND
Molecular cancer
CHD1L: a novel oncogene.
PMID 24359616
Abstract
Comprehensive sequencing efforts have revealed the genomic landscapes of common forms of human cancer and  - 140 driver genes have been identified, but not all of them have been extensively investigated. CHD1L (chromodomain helicase/ATPase DNA binding protein 1-like gene) or ALC1 (amplified in liver cancer 1) is a newly identified oncogene located at Chr1q21 and it is amplified in many solid tumors. Functional studies of CHD1L in hepatocellular carcinoma and other tumors strongly suggested that its oncogenic role in tumorigenesis is through unleashed cell proliferation, G1/S transition and inhibition of apoptosis. The underlying mechanisms of CHD1L activation may disrupt the cell death program via binding the apoptotic protein Nur77 or through activation of the AKT pathway by up-regulation of CHD1L-mediated target genes (e.g., ARHGEF9, SPOCK1 or TCTP). CHD1L is now considered to be a novel independent biomarker for progression, prognosis and survival in several solid tumors. The accumulated knowledge about its functions will provide a focus to search for targeted treatment in specific subtypes of tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV36364
Anti-CHD1L antibody produced in rabbit, IgG fraction of antiserum