EMAIL THIS PAGE TO A FRIEND
Cell
Probing DNA by 2-OG-dependent dioxygenase.
PMID 24360270
Abstract
TET-mediated 5-methyl cytosine (5mC) oxidation acts in epigenetic regulation, stem cell development, and cancer. Hu et al. now determine the crystal structure of the TET2 catalytic domain bound to DNA, shedding light on 5mC-DNA substrate recognition and the catalytic mechanism of 5mC oxidation.