EMAIL THIS PAGE TO A FRIEND
Neuron
Going mobile: AMPA receptors move synapse to synapse inxa0vivo.
PMID 24360537
Abstract
Plasticity models invoke the synaptic delivery of AMPARs, yet we know little about how receptors move inxa0vivo. In this issue of Neuron, Hoerndli etxa0al. (2013) show that lateral diffusion and kinesin-mediated transport move AMPARs between synapses inxa0vivo.