EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
4,4'-Unsymmetrically substituted 3,3'-biphenyl alpha helical proteomimetics as potential coactivator binding inhibitors.
PMID 24360824
Abstract
A series of unsymmetrically substituted biphenyl compounds was designed as alpha helical proteomimetics with the aim of inhibiting the binding of coactivator proteins to the nuclear hormone receptor coactivator binding domain. These compounds were synthesized in good overall yields in seven steps starting from 2-bromoanisole. The final products were evaluated using cotransfection reporter gene assays and mammalian two-hybrid competitive inhibition assays to demonstrate their effectiveness as competitive binding inhibitors. The results from this study indicate that these proteomimetics possess the ability to inhibit coactivator-receptor interactions, but via a mixed mode of inhibition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
159239
2-Bromoanisole, 97%
C7H7BrO