EMAIL THIS PAGE TO A FRIEND
American family physician
FPIN's clinical inquiries. Treatments for symptoms of the common cold.
PMID 24364554
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0361000 Ipratropium bromide impurity B, European Pharmacopoeia (EP) Reference Standard
C20H30BrNO3