EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Fluxes of Ca2+ and K+ are required for the listeriolysin O-dependent internalization pathway of Listeria monocytogenes.
PMID 24366251
Abstract
Listeria monocytogenes is responsible for the life-threatening food-borne disease listeriosis. This disease mainly affects elderly and immunocompromised individuals, causing bacteremia and meningoencephalitis. In pregnant women, L. monocytogenes infection leads to abortion and severe infection of the fetus or newborn. The L. monocytogenes intracellular life cycle is critical for pathogenesis. Previous studies have established that the major virulence factor of L. monocytogenes, the pore-forming toxin listeriolysin O (LLO), is sufficient to induce L. monocytogenes internalization into human epithelial cell lines. This internalization pathway strictly requires the formation of LLO pores in the plasma membrane and can be stimulated by the heterologous pore-forming toxin pneumolysin, suggesting that LLO acts nonspecifically by forming transmembrane pores. The present work tested the hypothesis that Ca2+ and K+ fluxes subsequent to perforation by LLO control L. monocytogenes internalization. We report that L. monocytogenes perforates the host cell plasma membrane in an LLO-dependent fashion at the early stage of invasion. In response to perforation, host cells undergo Ca2+ -dependent but K+ -independent resealing of their plasma membrane. In contrast to the plasma membrane resealing process, LLO-induced L. monocytogenes internalization requires both Ca2+ and K+ fluxes. Further linking ion fluxes to bacterial internalization, treating cells with a combination of Ca2+ and K+ ionophores but not with individual ionophores is sufficient to induce efficient internalization of large cargoes, such as 1-μm polystyrene beads and bacteria. We propose that LLO-induced L. monocytogenes internalization requires a Ca2+ - and K+ -dependent internalization pathway that is mechanistically distinct from the process of plasma membrane resealing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1520
Anti-Bovine Albumin antibody produced in rabbit, whole antiserum