EMAIL THIS PAGE TO A FRIEND
Journal of Chromatographic Science
Separation and quantitation of three acidic herbicide residues in tobacco and soil by dispersive solid-phase extraction and UPLC-MS/MS.
PMID 24366907