EMAIL THIS PAGE TO A FRIEND
Molecular immunology
The role of complement activation in the pathogenesis of Fuchs' dystrophy.
PMID 24368317
Abstract
Inflammation can be an etiologic factor of Fuchs' dystrophy according to previous studies. Our aim was to analyse the activation of the complement system in the aqueous humor in this pathological condition. 100 μl aqueous humor sample was taken during keratoplasty of 11 Fuchs' dystrophic patients and during phacoemulsification surgery of 18 control patients. The samples were mixed with EDTA and stored at -80 °C. Concentrations of C1rC1sC1Inh and C3bBbP complexes as markers of the activation of the classical and alternative complement pathways, respectively, were measured with ELISA method. The results of the patient group and the control group were compared with statistical analysis (non-parametric Mann Whitney test). Both the concentrations of C1rC1sC1Inh [4.3 (3.2-20.2)AU/ml] and of C3bBbP [15.3 (7.8-22.6)AU/ml] were significantly higher in the Fuchs' dystrophic group than in the control group [C1rC1sC1Inh: 0.0 (0.0-5.6)AU/ml, C3bBbP: 1.4 (0.0-7.8)AU/ml]. The median value is shown along with the (25% and 75% percentiles). Based on our results, the complement system may be activated both through the classical and alternative pathways in the aqueous humor of the patients with Fuchs' dystrophy.