EMAIL THIS PAGE TO A FRIEND
The journal of trauma and acute care surgery
Assessment of coagulopathy, endothelial injury, and inflammation after traumatic brain injury and hemorrhage in a porcine model.
PMID 24368352
Abstract
Traumatic brain injury (TBI) and hemorrhagic shock (HS) can be associated with coagulopathy and inflammation, but the mechanisms are poorly understood. We hypothesized that a combination of TBI and HS would disturb coagulation, damage the endothelium, and activate inflammatory and complement systems. A total of 33 swine were allocated to either TBI + HS (n = 27, TBI and volume-controlled 40% blood loss) or controls (n = 6, anesthesia and instrumentation). TBI + HS animals were left hypotensive (mean arterial pressure, 30-35 mm Hg) for 2 hours. Blood samples were drawn at baseline, 3 minutes and 15 minutes after injury, as well as following 2 hours of hypotension. Markers of coagulation, anticoagulation, endothelial activation/glycocalyx shedding, inflammation, complement, and sympathoadrenal function were measured. The TBI + HS group demonstrated an immediate (3 minutes after injury) activation of coagulation (prothrombin fragment 1 + 2, 289 ng/mL vs. 232 ng/mL, p = 0.03) and complement (C5a, 2.83 ng/mL vs. 2.05 ng/mL, p = 0.05). Shedding of the endothelial glycocalyx (syndecan 1) was evident 15 minutes after injury (851.0 ng/ml vs. 715.5 ng/ml, p = 0.03) while inflammation (tumor necrosis factor α [TNF-α], 81.1 pg/mL vs. 50.8 pg/mL, p = 0.03) and activation of the protein C system (activated protein C, 56.7 ng/mL vs. 26.1 ng/mL, p = 0.01) were evident following the 2-hour hypotension phase. The combination of TBI and shock results in an immediate activation of coagulation and complement systems with subsequent endothelial shedding, protein C activation, and inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1279
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
C6H7NaO6