EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
miR-338-3p suppresses gastric cancer progression through a PTEN-AKT axis by targeting P-REX2a.
PMID 24375644
Abstract
Results from recent studies suggest that aberrant microRNA expression is common in numerous cancers. Although miR-338-3p has been implicated in hepatocellular carcinoma, its role in gastric cancer is unknown. To this end, we report that miR-338-3p is downregulated in both gastric cancer tissue and cell lines. Forced expression of miR-338-3p inhibited cell proliferation and clonogenicity and induced a G1-S arrest as well as apoptosis in gastric cancer cells. Furthermore, P-Rex2a (PREX2) was identified as a direct target of miR-338-3p, and silencing P-Rex2a resulted in the same biologic effects of miR-338-3p expression in gastric cancer cells. Furthermore, both enforced expression of miR-338-3p or silencing of P-Rex2a resulted in activation of PTEN, leading to a decline in AKT phosphorylation. Also, miR-338-3p markedly inhibited the in vivo tumorigenicity in a nude mouse xenograft model system. These results demonstrate that miR-338-3p affects gastric cancer progression through PTEN-AKT signaling by targeting P-Rex2a in gastric cancer cells, which posits miR-338-3p as a novel strategy for gastric cancer treatment. miR-338-3p acts as a novel tumor suppressor that blocks the growth of gastric cancer cells through PTEN-PI3K signaling by targeting P-Rex2a.