EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Highly N2-selective coupling of 1,2,3-triazoles with indole and pyrrole.
PMID 24375713
Abstract
Hydrogen-bond mediated coupling of 1,2,3-triazoles to indoles and pyrroles results in N2 selective functionalization of the triazole moiety in moderate to excellent yields. The reaction was tolerant of un-, mono- and disubstituted triazoles and was applied to synthesize tryptophan derived fluorescent amino acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131709
Pyrrole, reagent grade, 98%
C4H5N