EMAIL THIS PAGE TO A FRIEND
Pediatric blood & cancer
Growth hormone treatment of patients with Fanconi anemia after hematopoietic cell transplantation.
PMID 24376097
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8648 Somatotropin (STH) from porcine pituitary