EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
Essential roles of PKA, iNOS, CD95/CD95L, and terminal caspases in suppression of eosinopoiesis by PGE2 and other cAMP-elevating agents.
PMID 24376378
Abstract
Up- and downregulation of eosinopoiesis control pulmonary eosinophilia in human asthma. In mice, eosinopoiesis is suppressed in vitro by prostaglandin E2 (PGE2) and in vivo by diethylcarbamazine, through a proapoptotic mechanism sequentially requiring inducible NO synthase (iNOS) and the ligand for death receptor CD95 (CD95L). We examined the roles of iNOS, cAMP-mediated signaling, caspases, and CD95L/CD95 in suppression of eosinopoiesis by PGE2 and other agents signaling through cAMP. Bone-marrow collected from BALB/c mice, or from iNOS-, CD95-, or CD95L-deficient mutants (and wild-type controls), was cultured with interleukin-5 (IL-5), alone or associated with PGE2, cAMP-inducing/mimetic agents, caspase inhibitor zVAD-fmk, iNOS inhibitor aminoguanidine, or combinations thereof, and eosinopoiesis was evaluated at various times. PGE2, added up to 24 hours of culture, dose-dependently suppressed eosinopoiesis, by inducing apoptosis. This effect was (a) paralleled by induction of iNOS in eosinophils; (b) duplicated by sodium nitroprusside, isoproterenol, and cAMP-inducing/mimetic agents; (c) prevented by protein kinase A inhibition. NO was produced through iNOS by dibutyryl-cAMP-stimulated bone-marrow. Overall, PGE2 and isoproterenol shared a requirement for four effector elements (iNOS, CD95L, CD95, and terminal caspases), which together define a pathway targeted by several soluble up- and downmodulators of eosinopoiesis, including drugs, mediators of inflammation, and cytokines.