EMAIL THIS PAGE TO A FRIEND
Journal of computer assisted tomography
Correlations between tumor stroma characters and dynamic enhanced MDCT findings in nodular pulmonary adenocarcinoma.
PMID 24378886
Abstract
This study aimed to evaluate correlations between tumor stroma characters and dynamic contrast-enhanced computed tomographic (CT) findings in nodular pulmonary adenocarcinoma. Thirty-three patients with nodular pulmonary adenocarcinoma underwent dynamic contrast-enhancement CT scan before surgery. CT findings include wash-in, wash-out, and distribution of enhancement. The proportion of invasive and noninvasive stroma in tumor was calculated. Invasive and noninvasive stroma proportion in tumor was correlated positively with wash-in and wash-out enhancement, respectively. Tumor stroma proliferation may explain the pathologic basis of CT dynamic enhancement and be a useful prognostic factor of pulmonary adenocarcinoma.