EMAIL THIS PAGE TO A FRIEND
Occupational health & safety (Waco, Tex.)
Exploring crystalline silica exposure.
PMID 24380193
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
421553
Silica
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si
541036
Silica, mesostructured, HMS (wormhole)
O2Si
643637
Silica, mesostructured, MSU-H (large pore 2D hexagonal)
O2Si
643645
Silica, mesostructured, MCM-41 type (hexagonal)
O2Si
637238
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
O2Si
637246
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
O2Si
701491
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, 99.99% trace metals basis
O2Si