EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Mechanism of drug failure in fusarium keratitis, 2004-2006.
PMID 24382083
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8986
Alexidine dihydrochloride, ≥95% (HPLC)
C26H56N10 · 2HCl