EMAIL THIS PAGE TO A FRIEND
Genes & nutrition
Lipocalin-2 (Lcn2) expression is mediated by maternal nutrition during the development of the fetal liver.
PMID 24382649
Abstract
The mechanisms by which maternal protein deficiency programs insulin action in the offspring are poorly understood. The interpretation of transcriptomics is complicated by homeostatic adaptations, for example, changes in amino acid metabolism, which are potentially unrelated to the programming mechanism. The fatty acid composition of the maternal diet modulates the programming of insulin action, offering a possible strategy to circumvent these complications. Fetal livers harvested on d21 of gestation from pregnant rats fed high-protein (18xa0% w/w) and low-protein (9xa0% w/w) diets prepared with either corn or soya oil were screened with rat genome microarrays. Although a low-protein maternal diet altered the abundance of more than one hundred mRNAs in the fetal liver, only 40 were changed by the fatty acid composition of the diet (Pxa0