EMAIL THIS PAGE TO A FRIEND
The American journal of medicine
Anaphylaxis challenges on the front line: perspectives from community medicine.
PMID 24384135
Abstract
This report reflects a discussion from the multidisciplinary Partnership for Anaphylaxis Round Table meeting, held in November 2012, in Dallas, Texas. Community medicine participants included John R. Bennett, MD, an internist who practiced in Cumming, Georgia, and whose patients were adults; Leonard Fromer, MD, a family practitioner in Los Angeles, California, who was the medical director of a network of 600 medical groups, including pediatricians, internists, and family physicians, and who in his previous practice treated children and adults, many of them with severe allergies; and Mary Lou Hayden, MS, RN, FNP-BC, AE-C, a nurse practitioner who treated adults in a university employee health clinic and in an allergy clinic in Charlottesville, Virginia, and whose prior practice focused on allergy and immunology in children and adults. This discussion was moderated by Dr Bennett. Participants provided their perspectives as primary care providers (PCPs) concerning anaphylaxis, which has become a major public health concern. The rising prevalence of severe allergies and incidence of anaphylaxis and other severe allergic reactions among children and adults is shifting more care to PCPs. This discussion provides insights into challenges faced by PCPs in treating patients at risk for anaphylaxis in the community setting and provides potential solutions to those challenges.