EMAIL THIS PAGE TO A FRIEND
Andrologia
Human leucocytes in asthenozoospermic patients: endothelial nitric oxide synthase expression.
PMID 24386917
Abstract
In a basic study at the Andrology Unit, Department of Clinical and Molecular Sciences, Polytechnic University of Marche, Ancona, Italy, we evaluated the pattern of mRNA endothelial nitric oxide synthase (eNOS) expression in human blood leucocytes isolated from normozoospermic fertile and asthenozoospermic infertile men to elucidate any pathogenic involvement in sperm cell motility. Forty infertile men with idiopathic asthenozoospermia and 45 normozoospermic fertile donors, age-matched, were included. Semen parameters were evaluated, and expression analysis of mRNA was performed in human leucocytes using reverse transcription polymerase chain reaction. Sperm volume, count, motility and morphology were determined, and eNOS expression and Western blotting analyses were performed. A positive correlation was observed between the concentrations of NO and the percentage of immotile spermatozoa. The mRNA of eNOS was more expressed in peripheral blood leucocytes isolated from asthenozoospermic infertile men versus those of fertile normozoospermic men (7.46 ± 0.38 versus 7.06 ± 0.56, P = 0.0355). A significant up-regulation of eNOS gene in peripheral blood leucocytes was 1.52-fold higher than that of fertile donors. It is concluded that eNOS expression and activity are enhanced in blood leucocytes in men with idiopathic asthenozoospermia.