EMAIL THIS PAGE TO A FRIEND
The Journal of allergy and clinical immunology
Prostaglandin D2 activates group 2 innate lymphoid cells through chemoattractant receptor-homologous molecule expressed on TH2 cells.
PMID 24388011
Abstract
Activation of the group 2 innate lymphoid cell (ILC2) population leads to production of the classical type 2 cytokines, thus promoting type 2 immunity. Chemoattractant receptor-homologous molecule expressed on TH2 cells (CRTH2), a receptor for prostaglandin D₂ (PGD₂), is expressed by human ILC2s. However, the function of CRTH2 in these cells is unclear. We sought to determine the role of PGD₂ and CRTH2 in human ILC2s and compare it with that of the established ILC2 activators IL-25 and IL-33. The effects of PGD₂, IL-25, and IL-33 on the cell migration, cytokine production, gene regulation, and receptor expression of ILC2s were measured with chemotaxis, ELISA, Luminex, flow cytometry, quantitative RT-PCR, and QuantiGene assays. The effects of PGD₂ under physiologic conditions were evaluated by using the supernatant from activated mast cells. PGD₂ binding to CRTH2 induced ILC2 migration and production of type 2 cytokines and many other cytokines. ILC2 activation through CRTH2 also upregulated the expression of IL-33 and IL-25 receptor subunits (ST2 and IL-17RA). The effects of PGD₂ on ILC2s could be mimicked by the supernatant from activated human mast cells and inhibited by a CRTH2 antagonist. PGD₂ is an important and potent activator of ILC2s through CRTH2 mediating strong proallergic inflammatory responses. Through IgE-mediated mast cell degranulation, these innate cells can also contribute to adaptive type 2 immunity; thus CRTH2 bridges the innate and adaptive pathways in human ILC2s.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5172
Prostaglandin D2, ≥95%, synthetic
C20H32O5