EMAIL THIS PAGE TO A FRIEND
Pediatric blood & cancer
Lack of association between MDM2 promoter SNP309 and clinical outcome in patients with neuroblastoma.
PMID 24391119
Abstract
While a polymorphism located within the promoter region of the MDM2 proto-oncogene, SNP309 (T > G), has previously been associated with increased risk and aggressiveness of neuroblastoma and other tumor entities, a protective effect has also been reported in certain other cancers. In this study, we evaluated the association of MDM2 SNP309 with outcome in 496 patients with neuroblastoma and its effect on MDM2 expression. No significant difference in overall or event-free survival was observed among patients with neuroblastoma with or without MDM2 SNP309. The presence of SNP309 does not affect MDM2 expression in neuroblastoma.
Related Materials