EMAIL THIS PAGE TO A FRIEND
Harvard heart letter : from Harvard Medical School
New alternatives to warfarin. New drugs may be best when starting treatment for atrial fibrillation, but don't switch if warfarin works for you.
PMID 24392500