EMAIL THIS PAGE TO A FRIEND
JPMA. The Journal of the Pakistan Medical Association
Use of a levonorgestrel-releasing intrauterine device in the treatment of adenomyosis associated heavy menstrual bleeding.
PMID 24392517
Abstract
To evaluate the effects of a levonorgestrel-releasing intrauterine device in the treatment of adenomyosis associated with heavy menstrual bleeding. The retrospective study was conducted at a tertiary referral hospital in Izmir, Turkey, and comprised data on adenomyosis patients who were implanted with a levonorgestrel-releasing intrauterine device for heavy menstrual bleeding between December 2004 and January 2008. After the insertion of the device, all patients were followed up by transvaginal ultrasonography and serum haemoglobin levels and menstrual patterns were determined at the 6th and 12th month. Data was analysed using SPSS 10. The mean age of the 42 women in the study was 43.2 +/- 0.8 years. At the sixth month, amenorrhoea, oligomenorrhoea, spotting and regular menstrual flow were 9.5% (n = 4), 7% (n = 3), 19% (n = 8), and 64% (n = 27), respectively. At the 12th month, the same parametres were 9.5% (n = 4), 7% (n = 3), 12% (n = 5), and 71% (n = 30), respectively. Haemoglobin levels had increased and endometrial thickness had decreased, and these differences were statistically significant (p < 0.001). The easy-to-use levonorgestrel-releasing intrauterine device can be added to the treatment options as a well-tolerated alternative in cases where a woman who has completed her fertility and does not request a hysterectomy has anaemia associated with adenomyosis.