EMAIL THIS PAGE TO A FRIEND
Gan to kagaku ryoho. Cancer & chemotherapy
[Treatment outcome of peptide vaccination for advanced colorectal cancer].
PMID 24393856
Abstract
Complementary DNA( cDNA) microarray technology coupled with laser microdissection has been used to identify human leukocyte antigen (HLA)-A24-restricted epitope peptides as potential targets for cancer vaccination in colorectal cancer patients. These antigenic peptides were derived from 2 different cancer-testis antigens, ring finger protein 43 (RNF43) and translocase of outer mitochondrial membrane 34( TOMM34). We conducted a clinical trial of colorectal cancer-specific peptide( RNF43, TOMM34) vaccines with uracil/tegafur( UFT)+Leucovorin( LV) for the treatment of advanced or recurrent colorectal cancer. The vaccinations were well tolerated without any serious adverse events. There were long-term survivors in the group showing cytotoxic T lymphocyte (CTL) responses against both RNF43 and TOMM34, as well as in the group showing CTL responses against either RNF43 or TOMM34. A new study has been planned to obtain more immunological responses. We started a clinical trial of vaccines against multiple peptides (RNF43, TOMM34, forkhead box protein M1 [FOXM1], maternal embryonic leucine zipper kinase [MELK], holliday junction recognition protein[HJURP], vascular endothelial growth factor receptor 1[VEGFR1], and VEGFR2) for the treatment of advanced or recurrent colorectal cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47612
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)
C20H21CaN7O7 · xH2O
F7878
Folinic acid calcium salt hydrate, analytical standard
C20H21CaN7O7 · xH2O