EMAIL THIS PAGE TO A FRIEND
Cell Cycle
Phospho-?Np63?/microRNA network modulates epigenetic regulatory enzymes in squamous cell carcinomas.
PMID 24394434
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-689
Anti-EZH2 Antibody, Upstate®, from rabbit