EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Improved permeability of acyclovir: optimization of mucoadhesive liposomes using the phospholipid vesicle-based permeation assay.
PMID 24395733