Iranian Red Crescent medical journal
The Effect of Unsaturated Fatty Acids on Molecular Markers of Cholesterol Homeostasis in THP-1 Macrophages.
PMID 24396573
Macrophages derived foam cells are key factors in the maladaptive immune and inflammatory response. The study of the cholesterol homeostasis and the molecular factor involved in these cells is very important in understanding the process of atherosclerosis and the mechanisms that prevent its occurrence. This experimental study investigated the effects of c9, t11-Conjugated Linoleic Acid (c9, t11-CLA). Alpha Linolenic Acid (LA), and Eicosapentaenoic Acid (EPA) on the PPARα and ACAT1 mRNA expression by Real time PCR and cholesterol homeostasis in THP-1 macrophages derived foam cells. Incubation of CLA, LA, EPA, and synthetic ligands did not prevent increasing the cellular total cholesterol (TC). Free cholesterol (FC) is increased by Sandoz58-035 (P = 0.024) and decreased by fatty acids and Wy14643 (Pirinixic acid) (P = 0.035). The pattern of distribution of %EC is similar to the EC pattern distribution. The ACAT1 mRNA expression was significantly increased by EPA (P = 0.009), but c9, t11- CLA, LA, Wy14643, and Sandoz58-035 had no significant effect on the mRNA level of ACAT1 expression compared to DMSO(Dimethyl sulfoxide). In comparison to the control of Wy14643, Sandoz58-035, c9 and t11-CLA, EPA increased the PPARα mRNA levels (P = 0.024, P = 0.041, P = 0.043, and P = 0.004, respectively), even though, LA had no significant effect on the PPARα mRNA expression (P = 0.489). Variations in the chemical structure of fatty acids can affect their physiological function.
Related Materials
Product #
Molecular Formula
Add to Cart
Sandoz 58-035, >98% (HPLC), powder