EMAIL THIS PAGE TO A FRIEND
Genome Biology
Identification of distinct miRNA target regulation between breast cancer molecular subtypes using AGO2-PAR-CLIP and patient datasets.
PMID 24398324
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-590
Anti-Ago 2 Antibody, clone, Upstate®, from rabbit