EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Synergistic interaction between fentanyl and bupivacaine given intrathecally for labor analgesia.
PMID 24398818
Abstract
Lipophilic opioids and local anesthetics are often given intrathecally in combination for labor analgesia. However, the nature of the pharmacologic interaction between these drugs has not been clearly elucidated in humans. Three hundred nulliparous women randomly received 1 of 30 different combinations of fentanyl and bupivacaine intrathecally using a combined spinal-epidural technique for analgesia in the first stage of labor. Visual analogue scale pain scores were recorded for 30 min. Response was defined by percentage decrease in pain score from baseline at 15 and 30 min. Dose-response curves for individual drugs were fitted to a hyperbolic dose-response model using nonlinear regression. The nature of the drug interaction was determined using dose equivalence methodology to compare observed effects of drug combinations with effects predicted by additivity. The derived dose-response models for individual drugs (doses in micrograms) at 15 min were: Effect = 100 × dose / (13.82 + dose) for fentanyl, and Effect = 100 × dose / (1,590 + dose) for bupivacaine. Combinations of fentanyl and bupivacaine produced greater effects than those predicted by additivity at 15 min (P < 0.001) and 30 min (P = 0.015) (mean differences, 9.1 [95% CI, 4.1-14.1] and 6.4 [95% CI, 1.2-11.5] units of the normalized response, respectively), indicating a synergistic interaction. The pharmacologic interaction between intrathecal fentanyl and bupivacaine is synergistic. Characterization and quantification of this interaction provide a theoretical basis and support for the clinical practice of combining intrathecal opioids and local anesthetics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1160000 Bupivacaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C18H28N2O · HCl