EMAIL THIS PAGE TO A FRIEND
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine
[Case report; a case of lung and skin nocardiosis in a non-immunocompromised patient].
PMID 24400549