EMAIL THIS PAGE TO A FRIEND
International journal of oncology
ROS-mediated JNK/p38-MAPK activation regulates Bax translocation in Sorafenib-induced apoptosis of EBV-transformed B cells.
PMID 24402682
Abstract
Sorafenib (SRF) is a multi-kinase inhibitor that has been shown to have antitumor activity against several types of cancers, but the effect of SRF on EBV-transformed B cells is unknown. We report that SRF can induce the apoptosis of EBV-transformed B cells through JNK/p38-MAPK activation. SRF triggered the generation of reactive oxygen species (ROS), translocation of Bax into the mitochondria, disruption of mitochondrial membrane potential, activation of caspase-9, caspase-3 and PARP, and subsequent apoptosis. Moreover, we found that SRF exposure activated the phosphorylation of JNK and p38-MAPK and suppressed the phosphorylation of PI3K-p85 and Akt. N-acetyl-l-cysteine (NAC) inhibited the activation of JNK and p38-MAPK. SP600125 and SB203580 blocked apoptosis and mitochondrial membrane disruption but did not affect ROS production after SRF treatment. These findings provide novel insights into the molecular mechanisms driving SRF-mediated cell death and suggest that SRF could be a potential therapeutic drug for the treatment of EBV-related malignant diseases.