EMAIL THIS PAGE TO A FRIEND
Catheterization and Cardiovascular Interventions : Official Journal of the Society For Cardiac Angiography & Interventions
Transcatheter treatment of tricuspid regurgitation by caval valve implantation--experimental evaluation of decellularized tissue valves in central venous position.
PMID 24403276