EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Effective catalytic disproportionation of aqueous H2O2 with di- and mono-nuclear manganese(II) complexes containing pyridine alcohol ligands.
PMID 24406400
Abstract
The two novel manganese(II) complexes with 2-hydroxymethylpyridine (2-CH2OHpy) {[Mn2(μ-Cl)2(2-CH2OHpy)4]Cl2·2H2O (1)} and 2-hydroxyethylpyridine (2-(CH2)2OHpy) {[Mn(2-(CH2)2OHpy)2(NCS)2] (2)} were synthesized and characterized by means of X-ray diffraction, IR, EPR, HF EPR spectroscopy, magnetic and TG/DTG data. The complexes show catalase-like activity in neutral aqueous solution since they were able to disproportionate H2O2 to harmless H2O and O2. Both complexes act as true catalysts since they reverted to their original form after depleting all the H2O2, as suggested by the operando resonant inelastic X-ray spectroscopy (RIXS) measurements.