EMAIL THIS PAGE TO A FRIEND
Archives of Dermatological Research
Oral administration of Bifidobacterium breve attenuates UV-induced barrier perturbation and oxidative stress in hairless mice skin.
PMID 24414333