EMAIL THIS PAGE TO A FRIEND
BMC Genomics
Characterization of Toxoplasma gondii subtelomeric-like regions: identification of a long-range compositional bias that is also associated with gene-poor regions.
PMID 24417889