EMAIL THIS PAGE TO A FRIEND
International Journal of Obesity
Reduced hepatic aquaporin-9 and glycerol permeability are related to insulin resistance in non-alcoholic fatty liver disease.
PMID 24418844