EMAIL THIS PAGE TO A FRIEND
Biotechnology Journal
Angiogenic/osteogenic response of BMMSCs on bone-derived scaffold: effect of hypoxia and role of PI3K/Akt-mediated VEGF-VEGFR pathway.
PMID 24421279
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300609
Anti-FLT4 antibody produced in rabbit, affinity isolated antibody
SAB4300356
Anti-KDR (Ab-1175) antibody produced in rabbit, affinity isolated antibody
SAB4300357
Anti-KDR (Ab-1214) antibody produced in rabbit, affinity isolated antibody
SAB4300358
Anti-KDR (Ab-951) antibody produced in rabbit, affinity isolated antibody
SAB2900238
Anti-KDR (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900239
Anti-KDR (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101239
Anti-KDR antibody produced in rabbit, affinity isolated antibody
V1014 Anti-Vascular Endothelial Growth Factor Receptor 2 antibody produced in goat, affinity isolated antibody, lyophilized powder
GW21181 Anti-VEGFR-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300657
Anti-VEGFR2 (Ab-1059) antibody produced in rabbit, affinity isolated antibody
SAB4501645
Anti-VEGFR2 antibody produced in rabbit, affinity isolated antibody
WH0002324M1
Monoclonal Anti-FLT4 antibody produced in mouse, clone 5B6, purified immunoglobulin, buffered aqueous solution
SAB1403833
Monoclonal Anti-FLT4 antibody produced in mouse, clone 5B5, purified immunoglobulin, buffered aqueous solution
SAB1409022
Monoclonal Anti-FLT4 antibody produced in mouse, clone 1C1, purified immunoglobulin, buffered aqueous solution
WH0003791M1
Monoclonal Anti-KDR antibody produced in mouse, clone 2A2, purified immunoglobulin, buffered aqueous solution
SAB1404004
Monoclonal Anti-KDR antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
V9134
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, clone KDR-1, ascites fluid
SAB1305566
MONOCLONAL ANTI-VEGFR3 antibody produced in mouse, clone 818CT12.1.1, IgG fraction of antiserum, buffered aqueous solution