EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Comparison of different preparation methods of tanshinoneporous silica solid dispersion].
PMID 24422390
Abstract
Porous silica was used as a carrier to prepare tanshinone solid dispersions (SDs). sThe effect of the spray drying method or the solvent method on the drug dissolution of SD was studied. The structure characteristics of SDs was analyzed by SEM, DSC,XPRD and FTIR. And in vitro dissolution was also investigated. The results showed that drugs were highly dispersed into SDs prepared by spray drying method and the solvent method in amorphous form. In addition, the results of the dissolution tested in vitro exhibited that the tanshinone I and tanshinone II A, accumulated dissolutions of SDs prepared using solvent achieved 80. 9% ,84. 6% and 86. 2% ,88. 7% within 45,60 min, respectively. And SDs prepared using spray-drying method were 92.7% ,95. 3% and 95. 8%, 97. 1% within 45,60 min, respectively. The tanshinone SDs were prepared successfully by spray drying method and solvent method. The SDs prepared by spray drying method was more conducive to improving the dissolution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
421553
Silica
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si
541036
Silica, mesostructured, HMS (wormhole)
O2Si
637238
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
O2Si
637246
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
O2Si
701491
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, 99.99% trace metals basis
O2Si
T5330
Tanshinone I, ≥98% (HPLC)
C18H12O3
80714
Tanshinone I, analytical standard
C18H12O3
T4952
Tanshinone IIA, ≥97% (HPLC)
C19H18O3
51704
Tanshinone IIA, analytical standard
C19H18O3